accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrMgBr

CAS RN 56812-60-3

IUPAC Name: bromo[(2-bromophenyl)methyl]magnesium
Synonyms: bromo[(2-bromophenyl)methyl]magnesium
Molecular Weight (g/mol): 274.24
Molecular Formula: C7H6Br2Mg
InChi Key: HDXZJAZTPVLPEX-UHFFFAOYSA-M
SMILES: Br[Mg]CC1=CC=CC=C1Br
Boiling Point 
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)