Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromobenzylmagnesium bromide, 0.25M solution in diethyl ether, AcroSeal™

Catalog No. AC431611000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC431611000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC431611000 Supplier Thermo Scientific Chemicals Supplier No. 431611000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 56812-60-3 | C7H6Br2Mg | 274.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56812-60-3 , 60-29-7
Molecular Formula C7H6Br2Mg
Molecular Weight (g/mol) 274.24
MDL Number MFCD01319903
InChI Key HDXZJAZTPVLPEX-UHFFFAOYSA-M
Synonym 2-bromobenzylmagnesium bromide, hdxzjaztpvlpex-uhfffaoysa-m, 2-bromobenzylmagnesium bromide solution, bromo 2-bromophenyl methyl magnesium, 2-bromobenzylmagnesium bromide 0.25 m in diethyl ether, 2-bromobenzylmagnesium bromide, 0.25m in diethyl ether, 2-bromobenzylmagnesium bromide solution, 0.25 m in diethyl ether, grignard reagent
PubChem CID 10858676
IUPAC Name bromo[(2-bromophenyl)methyl]magnesium
SMILES Br[Mg]CC1=CC=CC=C1Br

Specifications

CAS Min % 5.37, 87.47
CAS Max % 12.53, 94.63
Color Yellow to Brown
Density 0.7660g/mL
Boiling Point 34.6°C
Flash Point −40°C
Packaging AcroSeal™ Glass Bottle
Quantity 100 mL
Specific Gravity 0.766
Formula Weight 274.24
Physical Form Solution
Chemical Name or Material 2-Bromobenzylmagnesium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause drowsiness or dizziness.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Extremely flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.