accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5691-20-3

IUPAC Name: (1R,2S)-2-azaniumylcyclohexane-1-carboxylate
Synonyms: (1R,2S)-2-ammoniocyclohexane-1-carboxylate
Molecular Weight (g/mol): 143.19
Molecular Formula: C7H13NO2
InChi Key: USQHEVWOPJDAAX-RITPCOANSA-N
SMILES: [NH3+][C@H]1CCCC[C@H]1C([O-])=O
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)