CAS RN 57-37-4
Benactyzine Hydrochloride 98.0+%, TCI America™
CAS: 57-37-4 Molecular Formula: C20H26ClNO3 Molecular Weight (g/mol): 363.88 MDL Number: MFCD00012624 InChI Key: ZCEHOOLYWQBGQO-UHFFFAOYSA-N Synonym: benactyzine hydrochloride,neuroleptone,tranquilline,amitakon,benaktin,destendo,ibiotyzil,leucidil,nervacton,nervatil PubChem CID: 66448 IUPAC Name: hydrogen 2-(diethylamino)ethyl 2-hydroxy-2,2-diphenylacetate chloride SMILES: [H+].[Cl-].CCN(CC)CCOC(=O)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
MP Biomedicals, Inc Benactizine HCl, MP Biomedicals
CAS: 57-37-4 Molecular Formula: C20H26ClNO3 Molecular Weight (g/mol): 363.88 MDL Number: MFCD00012624 InChI Key: ZCEHOOLYWQBGQO-UHFFFAOYSA-N Synonym: benactyzine hydrochloride,neuroleptone,tranquilline,amitakon,benaktin,destendo,ibiotyzil,leucidil,nervacton,nervatil PubChem CID: 66448 IUPAC Name: hydrogen 2-(diethylamino)ethyl 2-hydroxy-2,2-diphenylacetate chloride SMILES: [H+].[Cl-].CCN(CC)CCOC(=O)C(O)(C1=CC=CC=C1)C1=CC=CC=C1