Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benactizine HCl, MP Biomedicals

Catalog No. ICN203578 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN203578 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN203578 Supplier MP Biomedicals, Inc Supplier No. 0520357810

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 57-37-4
Molecular Formula C20H26ClNO3
Molecular Weight (g/mol) 363.88
MDL Number MFCD00012624
InChI Key ZCEHOOLYWQBGQO-UHFFFAOYSA-N
Synonym benactyzine hydrochloride, neuroleptone, tranquilline, amitakon, benaktin, destendo, ibiotyzil, leucidil, nervacton, nervatil
PubChem CID 66448
IUPAC Name hydrogen 2-(diethylamino)ethyl 2-hydroxy-2,2-diphenylacetate chloride
SMILES [H+].[Cl-].CCN(CC)CCOC(=O)C(O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 10 g
Formula Weight 363.89
Chemical Name or Material Benactizine HCl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.