Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benactyzine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. B00535G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B00535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B00535G Supplier TCI America Supplier No. B00535G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 57-37-4
Molecular Formula C20H26ClNO3
Molecular Weight (g/mol) 363.88
MDL Number MFCD00012624
InChI Key ZCEHOOLYWQBGQO-UHFFFAOYSA-N
Synonym benactyzine hydrochloride, neuroleptone, tranquilline, amitakon, benaktin, destendo, ibiotyzil, leucidil, nervacton, nervatil
PubChem CID 66448
IUPAC Name hydrogen 2-(diethylamino)ethyl 2-hydroxy-2,2-diphenylacetate chloride
SMILES [H+].[Cl-].CCN(CC)CCOC(=O)C(O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 178°C
Color White
Quantity 5 g
UN Number 2811
Formula Weight 363.88
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benactyzine Hydrochloride
EINECSNumber (9)-1343
RTECSNumber DD2800000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.