accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHOONCH3(R)NCH3(S)H3C

CAS RN 57-47-6

IUPAC Name: (3aS,8aR)-1,3a,8-trimethyl-1H,2H,3H,3aH,8H,8aH-pyrrolo[2,3-b]indol-5-yl N-methylcarbamate
Synonyms: physostigmine isopto eserine fysostigmin physostigmine, salicylate (1:1) physostol eserine salicylate ezerin physostigmine preparation 毒扁豆碱 physostigmine (substance)
Molecular Weight (g/mol): 275.35
Molecular Formula: C15H21N3O2
InChi Key: PIJVFDBKTWXHHD-HIFRSBDPSA-N
SMILES: CNC(=O)OC1=CC=C2N(C)[C@H]3N(C)CC[C@@]3(C)C2=C1
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results