accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COONH(R)NHOOClOHO

CAS RN 57096-11-4

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-6-({[(2-chlorophenyl)methoxy]carbonyl}amino)hexanoic acid
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]-6-({[(2-chlorophenyl)methoxy]carbonyl}amino)hexanoic acid
Molecular Weight (g/mol): 414.88
Molecular Formula: C19H27ClN2O6
InChi Key: ATUMDPHEFWGCJF-GGYSOQFKNA-N
SMILES: CC(C)(C)OC(=O)N[C@H](CCCCNC(=O)OCC1=CC=CC=C1Cl)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)