accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNOONOO

CAS RN 573-56-8

IUPAC Name: 2,6-dinitrophenol
Synonyms: 2,6-dinitrophenol dinitro-2,6-phenol o-dinitrophenol β-dinitrophenol
Molecular Weight (g/mol): 184.11
Molecular Formula: C6H4N2O5
InChi Key: JCRIDWXIBSEOEG-UHFFFAOYSA-N
SMILES: OC1=C(C=CC=C1[N+]([O-])=O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results