accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 57322-42-6

IUPAC Name: 3-{2-[4-(phenylamino)naphthalen-1-yl]diazen-1-yl}naphthalene-2,6-disulfonic acid
Synonyms: 3-{2-[4-(phenylamino)naphthalen-1-yl]diazen-1-yl}naphthalene-2,6-disulfonic acid
Molecular Weight (g/mol): 533.57
Molecular Formula: C26H19N3O6S2
InChi Key: UMZJOYSJYIDKDN-UHFFFAOYSA-N
SMILES: OS(=O)(=O)C1=CC=C2C=C(C(=CC2=C1)N=NC1=C2C=CC=CC2=C(NC2=CC=CC=C2)C=C1)S(O)(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results