accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)HNCH3OOO

CAS RN 57357-56-9

IUPAC Name: (2R)-2-acetamidopentanoate
Synonyms: (2R)-2-acetamidopentanoate
Molecular Weight (g/mol): 158.18
Molecular Formula: C7H12NO3
InChi Key: BSYFPUSAWVWWDG-ZCFIWIBFSA-M
SMILES: CCC[C@@H](NC(C)=O)C([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results