accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3O

CAS RN 5736-89-0

IUPAC Name: 1-(4-butoxyphenyl)ethan-1-one
Synonyms: 1-(4-butoxyphenyl)ethanone
Molecular Weight (g/mol): 192.26
Molecular Formula: C12H16O2
InChi Key: YYLCJWIQUFHYMY-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=C(C=C1)C(C)=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)