Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Butoxyacetophenone 98.0+%, TCI America™
SDP

Catalog No. B15015G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B15015G Supplier TCI America Supplier No. B15015G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5736-89-0
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.258
MDL Number MFCD00027200
InChI Key YYLCJWIQUFHYMY-UHFFFAOYSA-N
Synonym 1-4-butoxyphenyl ethanone, 4'-butoxyacetophenone, 4-butoxyacetophenone, 1-4-butoxyphenyl ethan-1-one, ethanone, 1-4-butoxyphenyl, p-butoxyacetophenone, 4'-n-butoxy acetophenone, 1-acetyl-4-butoxybenzene, p-butoxyacetophenon, p-n butoxyacetophenone
PubChem CID 79814
IUPAC Name 1-(4-butoxyphenyl)ethanone
SMILES CCCCOC1=CC=C(C=C1)C(=O)C

Specifications

Color White
Quantity 5 g
Formula Weight 192.26
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 4′-Butoxyacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.