Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-n-Butoxyacetophenone, 98%, Thermo Scientific™

Catalog No. AAL0846814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0846814 25 g
AAL0846806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0846814 Supplier Thermo Scientific Chemicals Supplier No. L0846814

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5736-89-0
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.258
MDL Number MFCD00027200
InChI Key YYLCJWIQUFHYMY-UHFFFAOYSA-N
Synonym 1-4-butoxyphenyl ethanone, 4'-butoxyacetophenone, 4-butoxyacetophenone, 1-4-butoxyphenyl ethan-1-one, ethanone, 1-4-butoxyphenyl, p-butoxyacetophenone, 4'-n-butoxy acetophenone, 1-acetyl-4-butoxybenzene, p-butoxyacetophenon, p-n butoxyacetophenone
PubChem CID 79814
IUPAC Name 1-(4-butoxyphenyl)ethanone
SMILES CCCCOC1=CC=C(C=C1)C(=O)C

Specifications

Melting Point 25°C to 27°C
Density 1.011
Boiling Point 172°C to 173°C (18 mmHg)
Flash Point >110°C (230°F)
Odor Sweet
Refractive Index 1.527
Quantity 25 g
Beilstein 637894
Formula Weight 192.26
Percent Purity 98%
Chemical Name or Material 4'-n-Butoxyacetophenone
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 227-248-4
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.