accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONNCH3H3CO(E)

CAS RN 574013-66-4

IUPAC Name: methyl (2E)-3-[3-(N-{[4'-(dimethylamino)-[1,1'-biphenyl]-4-yl]methyl}cyclohexaneamido)phenyl]prop-2-enoate
Synonyms: fexaramine
Molecular Weight (g/mol): 496.65
Molecular Formula: C32H36N2O3
InChi Key: VLQTUNDJHLEFEQ-KGENOOAVSA-N
SMILES: COC(=O)\C=C\C1=CC=CC(=C1)N(CC1=CC=C(C=C1)C1=CC=C(C=C1)N(C)C)C(=O)C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)