accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3NNH3C

CAS RN 5744-40-1

IUPAC Name: ethyl 1,3-dimethyl-1H-pyrazole-5-carboxylate
Synonyms: ethyl 2,5-dimethylpyrazole-3-carboxylate
Molecular Weight (g/mol): 168.2
Molecular Formula: C8H12N2O2
InChi Key: ZYSGPOXVDOROJU-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(C)=NN1C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results