accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClClCl

CAS RN 57465-28-8

IUPAC Name: 3,3',4,4',5-pentachloro-1,1'-biphenyl
Synonyms: 3,3',4,4',5-pentachloro-1,1'-biphenyl
Molecular Weight (g/mol): 326.42
Molecular Formula: C12H5Cl5
InChi Key: REHONNLQRWTIFF-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1Cl)C1=CC(Cl)=C(Cl)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results