accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3

CAS RN 575-43-9

IUPAC Name: 1,6-dimethylnaphthalene
Synonyms: 1,6-dimethylnaphthalene
Molecular Weight (g/mol): 156.23
Molecular Formula: C12H12
InChi Key: CBMXCNPQDUJNHT-UHFFFAOYSA-N
SMILES: CC1=CC=C2C(C)=CC=CC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results