CAS RN 5754-35-8
IUPAC Name:
2-(1,3-dioxolan-2-yl)ethan-1-amine
Synonyms:
2-(1,3-dioxolan-2-yl)ethanamine
Molecular Weight (g/mol):
117.15
Molecular Formula:
C5H11NO2
InChi Key:
JNAQYWWCTUEVKR-UHFFFAOYSA-N
SMILES:
NCCC1OCCO1
1
–
1
of
1
results
1
2-(2-Aminoethyl)-1,3-dioxolane 98.0+%, TCI America™
CAS: 5754-35-8 Molecular Formula: C5H11NO2 Molecular Weight (g/mol): 117.15 MDL Number: MFCD00142756 InChI Key: JNAQYWWCTUEVKR-UHFFFAOYSA-N PubChem CID: 79827 IUPAC Name: 2-(1,3-dioxolan-2-yl)ethan-1-amine SMILES: NCCC1OCCO1