Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Aminoethyl)-1,3-dioxolane 98.0+%, TCI America™
SDP

Catalog No. A11765G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11765G 5 g
A11761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A11765G Supplier TCI America Supplier No. A11765G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5754-35-8
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00142756
InChI Key JNAQYWWCTUEVKR-UHFFFAOYSA-N
PubChem CID 79827
IUPAC Name 2-(1,3-dioxolan-2-yl)ethan-1-amine
SMILES NCCC1OCCO1

Specifications

Color Colorless
Boiling Point 75°C
Quantity 5 g
UN Number 2733
Formula Weight 117.15
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(2-Aminoethyl)-1,3-dioxolane
EINECSNumber (5)-0501
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.