accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONOCH3

CAS RN 57597-62-3

IUPAC Name: 3,3-dimethoxypropanenitrile
Synonyms: 3,3-dimethoxypropanenitrile
Molecular Weight (g/mol): 115.13
Molecular Formula: C5H9NO2
InChi Key: JYHSJQNYYLGMEI-UHFFFAOYSA-N
SMILES: COC(CC#N)OC
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)