Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Dimethoxy-propanenitrile 97%, Thermo Scientific™

Catalog No. AC133541000
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC133541000 100 mL
AC133540250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC133541000 Supplier Thermo Scientific Chemicals Supplier No. 133541000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57597-62-3
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00044797
InChI Key JYHSJQNYYLGMEI-UHFFFAOYSA-N
Synonym 3,3-dimethoxypropionitrile, propanenitrile, 3,3-dimethoxy, 3,3-dimethoxypropiononitrile, 3,3-dimethoxy propionitrile, 3,3-dimethoxy-propionitrile, 3,3-dimethoxy-propanenitrile, 3,3-dimethoxypropanitrile, acmc-209lz7, #, propanenitrile,3,3-dimethoxy
PubChem CID 93731
IUPAC Name 3,3-dimethoxypropanenitrile
SMILES COC(CC#N)OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.026g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Refractive Index 1.4130 to 1.4150 (20°C, 589nm)
Linear Formula (CH3O)2CHCH2CN
Quantity 100 mL
Formula Weight 115.13
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 3, 3-Dimethoxypropanenitrile, 97%
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not eat, drink or smoke when using this product.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 260-842-1
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.