accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOHNOOHNH2NHOHN

CAS RN 576-19-2

IUPAC Name: 2-amino-6-(5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanamido)hexanoic acid
Synonyms: 2-amino-6-(5-{2-oxo-hexahydrothieno[3,4-d]imidazol-4-yl}pentanamido)hexanoic acid
Molecular Weight (g/mol): 372.48
Molecular Formula: C16H28N4O4S
InChi Key: BAQMYDQNMFBZNA-UHFFFAOYNA-N
SMILES: NC(CCCCNC(=O)CCCCC1SCC2NC(=O)NC12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results