accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3C(S)O(S)(S)OOHOO

CAS RN 57710-57-3

IUPAC Name: (5aS,9S,9aS)-1-oxo-6-(propan-2-yl)-3,5a,6,7,8,9a-hexahydro-1H-spiro[2-benzoxepine-9,2'-oxirane]-4-carboxylic acid
Synonyms: (5aS,9S,9aS)-6-isopropyl-1-oxo-3,5a,6,7,8,9a-hexahydrospiro[2-benzoxepine-9,2'-oxirane]-4-carboxylic acid
Molecular Weight (g/mol): 280.32
Molecular Formula: C15H20O5
InChi Key: JESMSCGUTIEROV-NOFREYILSA-N
SMILES: CC(C)C1CC[C@@]2(CO2)[C@@H]2[C@@H]1C=C(COC2=O)C(O)=O
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results