Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptelidic Acid, MP Biomedicals™

Catalog No. MP21833712 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
0.25 mg
1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21833712 0.25 mg
MP218337101 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP21833712 Supplier MP Biomedicals, Inc Supplier No. 02183371.2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent, selective, irreversible GAPDH inhibitor, as well as a DNA polymerase inhibitor. The koningic acid molecule, produced by fungi that consume sweet potatoes, has been shown to curb the excessive glucose consumption in tumors exhibiting the Warburg effect and leaving healthy cells alone.

  • Glyceraldehyde 3-phosphate dehydrogenase (GAPDH), a key enzyme in carbohydrate metabolism, reversibly catalyzes the conversion of GAP to 1,3-bisphosphoglycerate and NAD+
  • A sesquiterpene lactone produced by the fungus T. koningii that was shown to have antibiotic activity against anaerobic bacteria such as Bacteroides
  • It acts as an irreversible inhibitor of GAPDH that binds to the cysteine-149 residue at the active site of the enzyme (Ki = 1.6 μM)
  • Can selectively induce apoptosis in high-glycolytic cancer cells by inhibiting the generation of ATP in the glycolytic pathway
  • Heptelidic acid is also a selective and competitive inhibitor of mammalian DNA polymerases β and λ as well as terminal deoxynucleotidyl transferase in family X of DNA polymerases (Kis range from 5.2-9.5 μM)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57710-57-3
Molecular Formula C15H20O5
Synonym Koningic Acid

Specifications

Quantity 0.25 mg
Color White
Purity >95
Molecular Weight (g/mol) 280.3
Chemical Name or Material Heptelidic Acid
Recommended Storage -20°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.