accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO(S)OOHO

CAS RN 578-86-9

IUPAC Name: (2S)-7-hydroxy-2-(4-hydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-4-one
Synonyms: 5-deoxyflavanone (2S)-liquiritigenin liquiritigenin 4',7-dihydroxyflavanone
Molecular Weight (g/mol): 256.26
Molecular Formula: C15H12O4
InChi Key: FURUXTVZLHCCNA-AWEZNQCLSA-N
SMILES: OC1=CC=C(C=C1)[C@@H]1CC(=O)C2=CC=C(O)C=C2O1
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)