accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CCH3NCH3NO

CAS RN 58-15-1

IUPAC Name: 4-(dimethylamino)-1,5-dimethyl-2-phenyl-2,3-dihydro-1H-pyrazol-3-one
Synonyms: aminophenazone aminopyrine aminopyrene mamallet-A netsusarin amidophen dipyrine aminopyrinum dimethylaminophenazon amidopyrinum
Molecular Weight (g/mol): 231.3
Molecular Formula: C13H17N3O
InChi Key: RMMXTBMQSGEXHJ-UHFFFAOYSA-N
SMILES: CN(C)C1=C(C)N(C)N(C1=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (7)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (4)