Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Dimethylaminoantipyrine, 97%

Catalog No. AC115741000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC115741000 100 g Plastic bottle
AC115740050 5 g Glass Bottle
AC115740250 25 g Plastic bottle
AC115740010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC115741000 Supplier Thermo Scientific Chemicals Supplier No. 115741000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 58-15-1 | C13H17N3O | 231.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-15-1
Molecular Formula C13H17N3O
Molecular Weight (g/mol) 231.30
MDL Number MFCD00003142
InChI Key RMMXTBMQSGEXHJ-UHFFFAOYSA-N
Synonym aminophenazone, amidopyrine, aminopyrine, 4-dimethylaminoantipyrine, dipyrine, amidazophen, amidophen, aminopyrin, pyramidon, amidophenazone
PubChem CID 6009
ChEBI CHEBI:160246
IUPAC Name 4-(dimethylamino)-1,5-dimethyl-2-phenylpyrazol-3-one
SMILES CN(C)C1=C(C)N(C)N(C1=O)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 105°C to 110°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Quantity 100 g
Beilstein 24,46
Merck Index 15,47
Solubility Information Solubility in water: 1g/18mL
Formula Weight 231.3
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 4-Dimethylaminoantipyrine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-365-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.