Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Dimethylamino)antipyrine, 98+%

Catalog No. AAB214360B
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB214360B 1000 g
AAB2143618 50 g
AAB2143630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB214360B Supplier Thermo Scientific Chemicals Supplier No. B214360B
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 58-15-1 | C13H17N3O | 231.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-15-1
Molecular Formula C13H17N3O
Molecular Weight (g/mol) 231.30
MDL Number MFCD00003142
InChI Key RMMXTBMQSGEXHJ-UHFFFAOYSA-N
Synonym aminophenazone, amidopyrine, aminopyrine, 4-dimethylaminoantipyrine, dipyrine, amidazophen, amidophen, aminopyrin, pyramidon, amidophenazone
PubChem CID 6009
ChEBI CHEBI:160246
IUPAC Name 4-(dimethylamino)-1,5-dimethyl-2-phenylpyrazol-3-one
SMILES CN(C)C1=C(C)N(C)N(C1=O)C1=CC=CC=C1

Specifications

Melting Point 105°C to 109°C
Odor Odorless
Quantity 1000 g
UN Number UN2811
Merck Index 14,474
Formula Weight 231.3
Percent Purity ≥98%
Chemical Name or Material 4-(Dimethylamino)antipyrine
Hazard Category H301-H312-H315-H319-H335
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-365-8
RTECSNumber CD2625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.