accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3NOHHOHO

CAS RN 58-56-0

IUPAC Name: hydrogen 4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol chloride
Synonyms: hydrogen pyridoxine chloride
Molecular Weight (g/mol): 205.64
Molecular Formula: C8H12ClNO3
InChi Key: ZUFQODAHGAHPFQ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=NC=C(CO)C(CO)=C1O
Molecular Weight (g/mol) 
  • (10)
  • (10)
Percent Purity 
  • (3)
  • (4)
  • (3)
  • (5)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (5)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Grade 
  • (1)
  • (2)
  • (3)
  • (3)
  • (4)
  • (5)
Physical Form 
  • (5)
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (13)
  • (8)
  • (1)
  • (4)
  • (8)
  • (3)
  • (5)
  • (2)
  • (10)
  • (10)
  • (3)
  • (4)
  • (3)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (5)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (4)
  • (5)
  • (5)
  • (3)
  • (4)
  • (4)