Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridoxine Hydrochloride, FCC, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18605991 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-991 1 kg Amber Glass Bottle
18-605-990 100 g Amber Glass Bottle
18-605-989 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-605-991 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P14591KG
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

P1459, 58-56-0, C8H11NO3·HCl

Spectrum™ Chemical Pyridoxine Hydrochloride, FCC , a form of vitamin B6, is used as a vitamin B6 dietary supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 58-56-0
Molecular Formula C8H12ClNO3
Molecular Weight (g/mol) 205.64
InChI Key ZUFQODAHGAHPFQ-UHFFFAOYSA-N
IUPAC Name hydrogen 4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol chloride
SMILES [H+].[Cl-].CC1=NC=C(CO)C(CO)=C1O

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 1 kg
Percent Purity 98 to 100.5%
Grade FCC
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Pyridoxine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.