accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONSHN(E/Z)

CAS RN 5800-50-0

IUPAC Name: 5-ethylidene-3-phenyl-2-sulfanylideneimidazolidin-4-one
Synonyms: 5-ethylidene-3-phenyl-2-sulfanylideneimidazolidin-4-one
Molecular Weight (g/mol): 218.27
Molecular Formula: C11H10N2OS
InChi Key: SXYSWGDIVRMEDE-UHFFFAOYSA-N
SMILES: CC=C1NC(=S)N(C1=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results