accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 58026-14-5

IUPAC Name: 4-(benzyloxy)-2-methoxybenzaldehyde
Synonyms: 4-(benzyloxy)-2-methoxybenzaldehyde
Molecular Weight (g/mol): 242.27
Molecular Formula: C15H14O3
InChi Key: HZLNIOUBXRIFML-UHFFFAOYSA-N
SMILES: COC1=C(C=O)C=CC(OCC2=CC=CC=C2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results