accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5807-14-7

IUPAC Name: 1H,2H,3H,4H,6H,7H,8H-[1,3]diazino[1,2-a]pyrimidine
Synonyms: triazabicyclodecene
Molecular Weight (g/mol): 139.2
Molecular Formula: C7H13N3
InChi Key: FVKFHMNJTHKMRX-UHFFFAOYSA-N
SMILES: C1CNC2=NCCCN2C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)