accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSNNN

CAS RN 5810-55-9

IUPAC Name: 6-{[2-(adamantan-1-yl)-2-oxoethyl]sulfanyl}-[2,3'-bipyridine]-5-carbonitrile
Synonyms: 6-{[2-(adamantan-1-yl)-2-oxoethyl]sulfanyl}-[2,3'-bipyridine]-5-carbonitrile
Molecular Weight (g/mol): 389.52
Molecular Formula: C23H23N3OS
InChi Key: RRELDCZNWXJLLG-UHFFFAOYSA-N
SMILES: O=C(CSC1=NC(=CC=C1C#N)C1=CC=CN=C1)C12CC3CC(CC(C3)C1)C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results