accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 58263-67-5

IUPAC Name: diphenyl [chloro(phenyl)methyl]phosphonate
Synonyms: diphenyl chloro(phenyl)methylphosphonate
Molecular Weight (g/mol): 358.76
Molecular Formula: C19H16ClO3P
InChi Key: LTRZPFCYQMGZGD-UHFFFAOYNA-N
SMILES: ClC(C1=CC=CC=C1)P(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results