CAS RN 583-91-5
IUPAC Name:
calcium bis(2-hydroxy-4-(methylsulfanyl)butanoate)
Synonyms:
calcium bis(2-hydroxy-4-(methylsulfanyl)butanoate)
Molecular Weight (g/mol):
338.45
Molecular Formula:
C10H18CaO6S2
InChi Key:
ABRVDWASZFDIEH-UHFFFAOYNA-L
SMILES:
[Ca++].CSCCC(O)C([O-])=O.CSCCC(O)C([O-])=O
1
–
1
of
1
results
1
2-Hydroxy-4-(methylthio)butyric Acid (65-72% in Water), TCI America™
CAS: 583-91-5 Molecular Formula: C10H18CaO6S2 Molecular Weight (g/mol): 338.45 MDL Number: MFCD00070490 InChI Key: ABRVDWASZFDIEH-UHFFFAOYNA-L PubChem CID: 11427 IUPAC Name: calcium bis(2-hydroxy-4-(methylsulfanyl)butanoate) SMILES: [Ca++].CSCCC(O)C([O-])=O.CSCCC(O)C([O-])=O