accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 58305-05-8

IUPAC Name: 2-(chloromethoxy)ethyl benzoate
Synonyms: 2-(chloromethoxy)ethyl benzoate
Molecular Weight (g/mol): 214.65
Molecular Formula: C10H11ClO3
InChi Key: HNQDBKBFYQVYLX-UHFFFAOYSA-N
SMILES: ClCOCCOC(=O)C1=CC=CC=C1
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results