Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoyloxyethylchloromethylether, Bal. Mainly Bis(benzoyloxyethoxy)methane (Tech.) 85%, Thermo Scientific™

Catalog No. AC243430250
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC243430250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC243430250 Supplier Thermo Scientific Chemicals Supplier No. 243430250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58305-05-8
Molecular Formula C10H11ClO3
Molecular Weight (g/mol) 214.645
InChI Key HNQDBKBFYQVYLX-UHFFFAOYSA-N
Synonym 2-chloromethoxy ethyl benzoate, 2-benzoyloxyethoxymethyl chloride, 2-benzoyloxyethoxymethyl-chloride, benzoyloxyethyl chloromethyl ether, 1-benzoyloxy-2-chloromethoxyethane, 2-benzoyloxy ethoxymethyl chloride, ethanol, 2-chloromethoxy-, benzoate, 1-benzoyloxy-2-chloromethoxy ethane, ethanol, 2-chloromethoxy-, 1-benzoate
PubChem CID 2724500
IUPAC Name 2-(chloromethoxy)ethyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OCCOCCl

Specifications

CAS Min % 80.0
CAS Max % 100.0
Quantity 25 mL
Formula Weight 214.65
Grade Technical
Chemical Name or Material Benzoyloxyethylchloromethylether, tech., 85%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.