accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 58359-53-8

IUPAC Name: 4-(2-phenyldiazen-1-yl)benzene-1-sulfonyl chloride
Synonyms: 4-(2-phenyldiazen-1-yl)benzenesulfonyl chloride
Molecular Weight (g/mol): 280.73
Molecular Formula: C12H9ClN2O2S
InChi Key: UEAZPVORQYDKHZ-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results