CAS RN 58430-94-7
IUPAC Name:
3,5,5-trimethylhexyl acetate
Synonyms:
trimethylhexyl acetate
Molecular Weight (g/mol):
186.3
Molecular Formula:
C11H22O2
InChi Key:
DGKXDLCVQSQVBC-UHFFFAOYNA-N
SMILES:
CC(CCOC(C)=O)CC(C)(C)C