accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHOHOH3C(E)

CAS RN 58493-49-5

IUPAC Name: (9E)-N-[(4-hydroxy-3-methoxyphenyl)methyl]octadec-9-enamide
Synonyms: (9E)-N-[(4-hydroxy-3-methoxyphenyl)methyl]octadec-9-enamide
Molecular Weight (g/mol): 417.63
Molecular Formula: C26H43NO3
InChi Key: OPZKBPQVWDSATI-ZHACJKMWSA-N
SMILES: CCCCCCCC\C=C\CCCCCCCC(=O)NCC1=CC=C(O)C(OC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results