accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3COCH3

CAS RN 588-43-2

IUPAC Name: 1-(dibutoxymethoxy)butane
Synonyms: 1-(dibutoxymethoxy)butane
Molecular Weight (g/mol): 232.36
Molecular Formula: C13H28O3
InChi Key: SGJBIFUEFLWXJY-UHFFFAOYSA-N
SMILES: CCCCOC(OCCCC)OCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results