Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributyl Orthoformate 95.0+%, TCI America™
SDP

Catalog No. O026925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O026925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O026925ML Supplier TCI America Supplier No. O026925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 588-43-2
Molecular Formula C13H28O3
Molecular Weight (g/mol) 232.364
MDL Number MFCD00015250
InChI Key SGJBIFUEFLWXJY-UHFFFAOYSA-N
Synonym tributyl orthoformate, 1-dibutoxymethoxy butane, tri-n-butyl orthoformate, tributoxymethane, orthoformic acid butyl ester, orthoformic acid tributyl ester, orthoformic acid, tributyl ester, butane, 1,1',1-methylidynetris oxy tris, 1,1',1-methylidynetris oxy tributane
PubChem CID 68521
IUPAC Name 1-(dibutoxymethoxy)butane
SMILES CCCCOC(OCCCC)OCCCC

Specifications

Color Colorless
Boiling Point 247°C
Quantity 25 mL
Formula Weight 232.36
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Tributyl Orthoformate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.