accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3C(S)(S)CH3OOCH2

CAS RN 5888-33-5

IUPAC Name: (1S,4S)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-yl prop-2-enoate
Synonyms: isobornyl acrylate
Molecular Weight (g/mol): 208.3
Molecular Formula: C13H20O2
InChi Key: PSGCQDPCAWOCSH-BOURZNODSA-N
SMILES: CC1(C)[C@H]2CC[C@]1(C)C(C2)OC(=O)C=C
Boiling Point 
  • (2)
  • (1)
  • (4)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)