Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobornyl Acrylate (stabilized with MEHQ) 90.0+%, TCI America™
SDP

Catalog No. I0638500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0638500G 500 g
I063825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0638500G Supplier TCI America Supplier No. I0638500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5888-33-5
Molecular Formula C13H20O2
Molecular Weight (g/mol) 208.30
MDL Number MFCD00080424
InChI Key PSGCQDPCAWOCSH-BOURZNODSA-N
Synonym isobornyl acrylate, acrylic acid isobornyl ester, 1,7,7-trimethyltricyclo 2.2.1 hepten-2-yl-2-propenoate
PubChem CID 122130333
IUPAC Name (1S,4S)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-yl prop-2-enoate
SMILES CC1(C)[C@H]2CC[C@]1(C)C(C2)OC(=O)C=C

Specifications

Color Yellow
Boiling Point 104°C
Quantity 500 g
UN Number 3082
Formula Weight 208.30
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobornyl Acrylate (stabilized with MEHQ)
RTECSNumber UD3940000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.