CAS RN 58880-37-8
IUPAC Name:
1-(4-chlorophenyl)cyclohexane-1-carboxylate
Synonyms:
1-(4-chlorophenyl)cyclohexane-1-carboxylate
Molecular Weight (g/mol):
237.7
Molecular Formula:
C13H14ClO2
InChi Key:
UPNXUJXIIZGXLQ-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1(CCCCC1)C1=CC=C(Cl)C=C1
1
–
1
of
1
results
1
1-(4-Chlorophenyl)cyclohexane-1-carboxylic acid, 95%
CAS: 58880-37-8 Molecular Formula: C13H14ClO2 Molecular Weight (g/mol): 237.70 MDL Number: MFCD00019350 InChI Key: UPNXUJXIIZGXLQ-UHFFFAOYSA-M Synonym: 1-4-chlorophenyl cyclohexanecarboxylic acid,1-4-chlorophenyl cyclohexane-1-carboxylic acid,1-4-chlorophenyl-1-cyclohexanecarboxylic acid,1-4-chlorophenyl-1-cyclohexane-carboxylic acid,cyclohexanecarboxylic acid, 1-4-chlorophenyl,acmc-1cuiq,4-chlorophenylcyclohexanecarboxylic acid,4-chlorophenyl cyclohexanecarboxylic acid,1-4-chlorophenyl cyclohexanecarboxylicacid PubChem CID: 100873 IUPAC Name: 1-(4-chlorophenyl)cyclohexane-1-carboxylic acid SMILES: [O-]C(=O)C1(CCCCC1)C1=CC=C(Cl)C=C1