CAS RN 58895-64-0
IUPAC Name:
hydrogen (1S,5R,13S,17S)-4-(cyclopropylmethyl)-14-methylidene-12-oxa-4-azapentacyclo[9.6.1.0¹,¹³.0⁵,¹⁷.0⁷,¹⁸]octadeca-7,9,11(18)-triene-10,17-diol chloride
Synonyms:
hydrogen nalmefene chloride
Molecular Weight (g/mol):
375.89
Molecular Formula:
C21H26ClNO3
InChi Key:
GYWMRGWFQPSQLK-OPHZJPRHSA-N
SMILES:
[H+].[Cl-].OC1=CC=C2C[C@H]3N(CC4CC4)CC[C@@]45[C@@H](OC1=C24)C(=C)CC[C@@]35O
1
–
1
of
1
results
1
R&D Systems™ Nalmefene hydrochloride
CAS: 58895-64-0 Molecular Formula: C21H26ClNO3