accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 58932-13-1

H3CON

CAS RN 58932-13-1

IUPAC Name: 4'-(nonyloxy)-[1,1'-biphenyl]-4-carbonitrile
Synonyms: 4'-(nonyloxy)-[1,1'-biphenyl]-4-carbonitrile
Molecular Weight (g/mol): 321.46
Molecular Formula: C22H27NO
InChi Key: NJQLLUIPSJLJHY-UHFFFAOYSA-N
SMILES: CCCCCCCCCOC1=CC=C(C=C1)C1=CC=C(C=C1)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results