CAS RN 5896-54-8
IUPAC Name:
sodium pentadecane-1-sulfonate
Synonyms:
sodium pentadecane-1-sulfonate
Molecular Weight (g/mol):
314.46
Molecular Formula:
C15H31NaO3S
InChi Key:
PLQISZLZPSPBDP-UHFFFAOYSA-M
SMILES:
[Na+].CCCCCCCCCCCCCCCS([O-])(=O)=O
1
–
1
of
1
results
1
Sodium 1-Pentadecanesulfonate, TCI America™
CAS: 5896-54-8 Molecular Formula: C15H31NaO3S Molecular Weight (g/mol): 314.46 MDL Number: MFCD00059838 InChI Key: PLQISZLZPSPBDP-UHFFFAOYSA-M Synonym: 1-Pentadecanesulfonic Acid Sodium Salt PubChem CID: 23670835 IUPAC Name: sodium pentadecane-1-sulfonate SMILES: [Na+].CCCCCCCCCCCCCCCS([O-])(=O)=O